Assay Details
Target Gene Details
Entrez Gene ID: | 259217 |
Gene Name: | heat shock protein family A (Hsp70) member 12A |
Gene Aliases: |
- |
Location: |
Chr.10:116671192-116851241 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA12A | NM_025015.2 | 6 | 712 | NP_079291.2 |
| XM_005269672.3 | 7 | 862 | XP_005269729.1 | |
| XM_005269673.4 | 7 | 1806 | XP_005269730.1 | |
| XM_011539579.2 | 8 | 923 | XP_011537881.1 | |
| XM_011539580.2 | 7 | 868 | XP_011537882.1 | |
| XM_017016032.1 | 4 | 434 | XP_016871521.1 | |
| AB007877.1 | 6 | 712 | ||
| AK023936.1 | 1 | 747 | ||
| AK294672.1 | 5 | 526 | ||
| AK294884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624663 | Chr.10:116687516 - 116731008 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map