Assay Details
Target Gene Details
Entrez Gene ID: | 81621 |
Gene Name: | Kazal type serine peptidase inhibitor domain 1 |
Gene Aliases: |
BONO1, FKSG28, FKSG40, IGFBP-rP10 |
Location: |
Chr.10:101061141-101068141 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAZALD1 | NM_001319303.1 | 6 | 2694 | NP_001306232.1 |
| NR_135067.1 | 5 | 2077 | ||
| NR_135068.1 | 2 | 1026 | ||
| XM_017016714.1 | 5 | 1718 | XP_016872203.1 | |
| XM_017016715.1 | 5 | 1695 | XP_016872204.1 | |
| AF333487.1 | 3 | 819 | AAG50291.1 | |
| AK172864.1 | 7 | 2605 | ||
| AY014271.1 | 3 | 819 | AAG50354.1 | |
| BU737294.1 | 1 | 136 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map