Assay Details
Target Gene Details
Entrez Gene ID: | 104355295 |
Gene Name: | SGMS1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:50624951-50631636 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGMS1-AS1 | NR_126411.1 | |||
| DA202321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891809 | Chr.10:50242444 - 50647850 on Build GRCh38 | Gain |
|
| nsv1051087 | Chr.10:49977683 - 50687535 on Build GRCh38 | Loss |
|
| nsv469522 | Chr.10:50516804 - 50687829 on Build GRCh38 | Loss |
|

Genomic Map