Assay Details
Target Gene Details
Entrez Gene ID: | 55662 |
Gene Name: | hypoxia inducible factor 1 alpha subunit inhibitor |
Gene Aliases: |
FIH1 |
Location: |
Chr.10:100535884-100553924 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIF1AN | NM_017902.2 | 8 | 6387 | NP_060372.2 |
| XM_011539940.2 | 8 | 6398 | XP_011538242.1 | |
| XM_011539941.1 | 8 | 6265 | XP_011538243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv277e212 | Chr.10:100550193 - 100558814 on Build GRCh38 | Loss |
|
| nsv831963 | Chr.10:100458231 - 100678922 on Build GRCh38 | Loss |
|
| nsv831962 | Chr.10:100406551 - 100593251 on Build GRCh38 | Gain |
|
| esv2759779 | Chr.10:100392093 - 100805979 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map