Assay Details
Target Gene Details
Entrez Gene ID: | 404636 |
Gene Name: | family with sequence similarity 45 member A |
Gene Aliases: |
- |
Location: |
Chr.10:119104065-119137984 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552214 | Chr.10:119100409 - 119209705 on Build GRCh38 | Loss |
|
| nsv1041093 | Chr.10:118689033 - 119198373 on Build GRCh38 | Gain |
|

Genomic Map