Assay Details
Target Gene Details
Entrez Gene ID: | 100130887 |
Gene Name: | ATE1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121928312-121951965 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATE1-AS1 | NR_120495.1 | 2 | 567 | |
| AI634505.1 | ||||
| BF434328.1 | ||||
| BF513344.1 | 1 | 401 |

Genomic Map