Assay Details
Target Gene Details
Entrez Gene ID: | 90550 |
Gene Name: | mitochondrial calcium uniporter |
Gene Aliases: |
C10orf42, CCDC109A |
Location: |
Chr.10:72692131-72887694 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCU | NM_001270679.1 | NP_001257608.1 | ||
| NM_001270680.1 | NP_001257609.1 | |||
| NM_138357.2 | NP_612366.1 | |||
| NR_073062.1 | ||||
| AK075439.1 | ||||
| AK128016.1 | ||||
| AK301475.1 | ||||
| AK310455.1 | ||||
| AK311133.1 | ||||
| AK315519.1 | ||||
| BC010682.1 | AAH10682.1 | |||
| BC034235.1 | AAH34235.1 | |||
| HY146050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759767 | Chr.10:72675880 - 72847362 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map