Assay Details
Target Gene Details
Entrez Gene ID: | 439949 |
Gene Name: | PRKCQ antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:6580425-6585361 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCQ-AS1 | NR_036502.1 | 1 | 411 | |
| NR_036503.1 | 1 | 411 | ||
| AY007155.1 | 1 | 297 | ||
| CN429636.1 | 1 | 271 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047309 | Chr.10:6453621 - 6672098 on Build GRCh38 | Gain |
|
| nsv5698 | Chr.10:6564860 - 6590481 on Build GRCh38 | Insertion |
|
| nsv549912 | Chr.10:6525013 - 6587263 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114192161] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map