Assay Details
Target Gene Details
Entrez Gene ID: | 100132116 |
Gene Name: | ACTA2 antisense RNA 1 |
Gene Aliases: |
ZXF1, uc001kfo.1 |
Location: |
Chr.10:88932684-88939974 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA2-AS1 | NR_125373.1 | 2 | 377 | |
| AK093340.1 | 2 | 377 | ||
| AX748062.1 | 2 | 377 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain |
|
| nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain |
|
| esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|

Genomic Map