Assay Details
Target Gene Details
Entrez Gene ID: | 283045 |
Gene Name: | uncharacterized LOC283045 |
Gene Aliases: |
- |
Location: |
Chr.10:62339588-62375127 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC283045 | NR_104162.1 | |||
| AK095019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8681 | Chr.10:61896798 - 62671529 on Build GRCh38 | Gain |
|
| esv2761600 | Chr.10:62274926 - 62368374 on Build GRCh38 | Gain |
|
| dgv879n100 | Chr.10:62274914 - 62368374 on Build GRCh38 | Gain |
|
| esv34099 | Chr.10:62021186 - 62427805 on Build GRCh38 | Loss |
|

Genomic Map