Assay Details
Target Gene Details
Entrez Gene ID: | 643236 |
Gene Name: | transmembrane protein 72 |
Gene Aliases: |
C10orf127, KSP37 |
Location: |
Chr.10:44911307-44937010 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM72 | NM_001123376.2 | NP_001116848.1 | ||
| XM_011540071.2 | XP_011538373.1 | |||
| AB235418.1 | ||||
| BC127789.1 | ||||
| BC127790.1 |
Target Gene Details
Entrez Gene ID: | 220980 |
Gene Name: | TMEM72 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:44811024-44959689 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM72-AS1 | NR_033842.1 | |||
| AK056518.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517161 | Chr.10:44715242 - 44920808 on Build GRCh38 | Gain+Loss |
|
| nsv550714 | Chr.10:44912563 - 44951343 on Build GRCh38 | Gain |
|
| nsv831846 | Chr.10:44826788 - 45060922 on Build GRCh38 | Loss |
|
| esv2678805 | Chr.10:44915230 - 44919047 on Build GRCh38 | Deletion |
|
| nsv831845 | Chr.10:44804820 - 44964947 on Build GRCh38 | Gain |
|

Genomic Map