Assay Details
Target Gene Details
Entrez Gene ID: | 54906 |
Gene Name: | family with sequence similarity 208 member B |
Gene Aliases: |
C10orf18, bA318E3.2 |
Location: |
Chr.10:5684759-5763740 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522415 | Chr.10:5679690 - 5726530 on Build GRCh38 | Gain |
|
| nsv1050663 | Chr.10:5696016 - 5743955 on Build GRCh38 | Loss |
|
| dgv36n21 | Chr.10:5711774 - 5726530 on Build GRCh38 | Loss |
|
| dgv1e49 | Chr.10:5684858 - 6183224 on Build GRCh38 | Loss |
|
| nsv528504 | Chr.10:5697091 - 5712648 on Build GRCh38 | Gain |
|
| esv2759729 | Chr.10:5515061 - 5762817 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79175035] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map