Assay Details
Target Gene Details
Entrez Gene ID: | 92565 |
Gene Name: | fibronectin type III and ankyrin repeat domains 1 |
Gene Aliases: |
HSD13 |
Location: |
Chr.10:125896539-126009599 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANK1 | NM_145235.3 | NP_660278.3 | ||
| XM_005270280.3 | XP_005270337.1 | |||
| XM_011540349.2 | XP_011538651.1 | |||
| XM_011540350.2 | XP_011538652.1 | |||
| XM_011540360.2 | XP_011538662.1 | |||
| XM_011540363.2 | XP_011538665.1 | |||
| XM_017016917.1 | XP_016872406.1 | |||
| XM_017016919.1 | XP_016872408.1 | |||
| XM_017016920.1 | XP_016872409.1 | |||
| XM_017016921.1 | XP_016872410.1 | |||
| XM_017016922.1 | XP_016872411.1 | |||
| XM_017016923.1 | XP_016872412.1 | |||
| XM_017016924.1 | XP_016872413.1 | |||
| XM_017016925.1 | XP_016872414.1 | |||
| AK304407.1 | ||||
| AY358154.1 | AAQ88521.1 | |||
| BC024189.2 | AAH24189.2 | |||
| BQ225468.1 | ||||
| CR627249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761638 | Chr.10:125892202 - 126013242 on Build GRCh38 | Loss |
|
| dgv531n106 | Chr.10:125885831 - 125918431 on Build GRCh38 | Duplication |
|
| nsv951871 | Chr.10:125885932 - 125922631 on Build GRCh38 | Duplication |
|
| nsv1077324 | Chr.10:125886130 - 125900231 on Build GRCh38 | Duplication |
|
| dgv521e59 | Chr.10:125885593 - 125928391 on Build GRCh38 | Duplication |
|
| nsv8738 | Chr.10:125883224 - 125928907 on Build GRCh38 | Loss |
|
| esv2623784 | Chr.10:125886728 - 125906918 on Build GRCh38 | Gain |
|
| dgv179n67 | Chr.10:125884800 - 125928892 on Build GRCh38 | Loss |
|
| nsv948223 | Chr.10:125886503 - 125915974 on Build GRCh38 | Duplication |
|
| esv22980 | Chr.10:125888760 - 125902665 on Build GRCh38 | Gain |
|
| nsv1148754 | Chr.10:125887930 - 125925531 on Build GRCh38 | Duplication |
|
| esv2759788 | Chr.10:125765331 - 126098133 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map