Assay Details
Target Gene Details
Entrez Gene ID: | 100128511 |
Gene Name: | NEBL antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:21173990-21174923 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEBL-AS1 | NR_046283.1 | |||
| NR_046284.1 | ||||
| AB371527.1 | ||||
| AW204239.1 | ||||
| BX091902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831806 | Chr.10:21084709 - 21230421 on Build GRCh38 | Gain |
|
| nsv508573 | Chr.10:21168076 - 21236717 on Build GRCh38 | Deletion |
|

Genomic Map