Assay Details
Target Gene Details
Entrez Gene ID: | 80013 |
Gene Name: | family with sequence similarity 188 member A |
Gene Aliases: |
C10orf97, CARP, DERP5, MST126, MSTP126, my042 |
Location: |
Chr.10:15778169-15860533 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831795 | Chr.10:15683863 - 15832969 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147208753] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map