Assay Details
Target Gene Details
Entrez Gene ID: | 693189 |
Gene Name: | microRNA 604 |
Gene Aliases: |
MIRN604, hsa-mir-604 |
Location: |
Chr.10:29545004-29545097 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR604 | NR_030335.1 |
Target Gene Details
Entrez Gene ID: | 6840 |
Gene Name: | supervillin |
Gene Aliases: |
- |
Location: |
Chr.10:29457338-29737001 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038080 | Chr.10:29361590 - 29565510 on Build GRCh38 | Gain |
|
| nsv831819 | Chr.10:29514811 - 29681606 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map