Assay Details
Target Gene Details
Entrez Gene ID: | 340784 |
Gene Name: | H6 family homeobox 3 |
Gene Aliases: |
NKX-5.1, NKX5.1, Nkx5-1 |
Location: |
Chr.10:123136051-123137731 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMX3 | NM_001105574.1 | 2 | 518 | NP_001099044.1 |
| AW088771.1 |
More Information
Set Membership: |
|

Genomic Map