Assay Details
Target Gene Details
Entrez Gene ID: | 220906 |
Gene Name: | WAC antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.10:28519917-28532354 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WAC-AS1 | NR_033805.1 | 1 | 115 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734606 | Chr.10:27447765 - 29432048 on Build GRCh38 | Deletion |
|

Genomic Map