Assay Details
Target Gene Details
Entrez Gene ID: | 56 |
Gene Name: | acrosomal vesicle protein 1 |
Gene Aliases: |
D11S4365, SP-10, SPACA2 |
Location: |
Chr.11:125672334-125680898 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACRV1 | NM_001612.5 | NP_001603.1 | ||
| NM_020069.4 | NP_064454.1 | |||
| NM_020107.4 | NP_064492.1 | |||
| NM_020108.4 | NP_064493.1 | |||
| AB451476.1 | ||||
| AK223335.1 | BAD97055.1 | |||
| BC014588.2 | AAH14588.1 | |||
| BG720361.1 | ||||
| BI826178.1 | ||||
| M82967.1 | AAA36625.1 | |||
| M82968.1 | AAA36626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map