Assay Details
Target Gene Details
Entrez Gene ID: | 100129827 |
Gene Name: | MRVI1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:10541236-10599932 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1-AS1 | NR_034093.2 | 1 | 215 | |
| NR_034094.2 | 1 | 215 | ||
| NR_046374.1 | 1 | 215 | ||
| NR_046375.1 | 1 | 215 | ||
| AI075905.1 | 3 | 457 | ||
| BC028123.1 | 1 | 179 | AAH28123.1 | |
| BI520309.1 | 1 | 154 | ||
| BU569149.1 | 1 | 142 | ||
| BU570065.1 | 1 | 123 | ||
| DB071465.1 | 1 | 218 | ||
| DB454301.1 | 1 | 168 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044011 | Chr.11:10507806 - 10542903 on Build GRCh38 | Gain |
|
| nsv7675 | Chr.11:10532842 - 10565476 on Build GRCh38 | Insertion |
|

Genomic Map