Assay Details
Target Gene Details
Entrez Gene ID: | 170689 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 15 |
Gene Aliases: |
- |
Location: |
Chr.11:130448876-130478215 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS15 | NM_139055.3 | NP_620686.1 | ||
| XM_005271419.4 | XP_005271476.1 | |||
| AJ315733.1 | CAC86014.1 | |||
| BC109114.1 | AAI09115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759871 | Chr.11:130099327 - 130472822 on Build GRCh38 | Loss |
|
| nsv508660 | Chr.11:130397628 - 130503590 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115841719] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map