Assay Details
Target Gene Details
Entrez Gene ID: | 1315 |
Gene Name: | coatomer protein complex subunit beta 1 |
Gene Aliases: |
COPB |
Location: |
Chr.11:14457503-14499895 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPB1 | NM_001144061.1 | NP_001137533.1 | ||
| NM_001144062.1 | NP_001137534.1 | |||
| NM_016451.4 | NP_057535.1 | |||
| AF084457.1 | AAD41240.1 | |||
| AF111807.1 | AAL39009.1 | |||
| AK001203.1 | ||||
| AL136593.1 | CAB66528.1 | |||
| BC037280.2 | AAH37280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32960 | Chr.11:14453542 - 14490616 on Build GRCh38 | Loss |
|
| nsv951311 | Chr.11:14466055 - 14470654 on Build GRCh38 | Duplication |
|
| esv3578250 | Chr.11:14350967 - 14976647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map