Assay Details
Target Gene Details
Entrez Gene ID: | 1111 |
Gene Name: | checkpoint kinase 1 |
Gene Aliases: |
CHK1 |
Location: |
Chr.11:125624910-125676259 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHEK1 | NM_001114121.2 | 1 | 589 | NP_001107593.1 |
| NM_001114122.2 | 1 | 589 | NP_001107594.1 | |
| NM_001244846.1 | 1 | 589 | NP_001231775.1 | |
| NR_045204.1 | 1 | 589 | ||
| XM_011542560.2 | 1 | 815 | XP_011540862.1 | |
| XM_011542561.2 | 1 | 815 | XP_011540863.1 | |
| AK300082.1 | ||||
| BX419129.2 | 1 | 143 | ||
| CA868104.1 | 1 | 156 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map