Assay Details
Target Gene Details
Entrez Gene ID: | 54766 |
Gene Name: | BTG anti-proliferation factor 4 |
Gene Aliases: |
APRO3, PC3B |
Location: |
Chr.11:111383828-111512388 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTG4 | XM_011542876.2 | XP_011541178.1 | ||
| XM_011542878.2 | XP_011541180.1 |
Target Gene Details
Entrez Gene ID: | 100132078 |
Gene Name: | uncharacterized LOC100132078 |
Gene Aliases: |
- |
Location: |
Chr.11:111414242-111418186 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100132078 | NR_046085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054465 | Chr.11:111402183 - 111438447 on Build GRCh38 | Gain |
|
| nsv832268 | Chr.11:111311611 - 111523324 on Build GRCh38 | Loss |
|
| nsv1053752 | Chr.11:111413921 - 111440592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map