Assay Details
Target Gene Details
Entrez Gene ID: | 79659 |
Gene Name: | dynein cytoplasmic 2 heavy chain 1 |
Gene Aliases: |
ATD3, DHC1b, DHC2, DNCH2, DYH1B, SRPS2B, SRTD3, hdhc11 |
Location: |
Chr.11:103109431-103479863 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNC2H1 | NM_001080463.1 | 2 | 390 | NP_001073932.1 |
| NM_001377.2 | 2 | 390 | NP_001368.2 | |
| XM_006718903.2 | 2 | 390 | XP_006718966.1 | |
| XM_017018291.1 | 2 | 390 | XP_016873780.1 | |
| XM_017018293.1 | 2 | 390 | XP_016873782.1 | |
| AB231766.1 | 2 | 299 | BAE17138.1 | |
| AB290167.1 | 2 | 444 | ||
| AK125524.1 | 2 | 390 | BAC86194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468855 | Chr.11:103066950 - 103185951 on Build GRCh38 | Loss |
|
| esv3627546 | Chr.11:103065903 - 103212201 on Build GRCh38 | Gain |
|
| nsv1045952 | Chr.11:103067310 - 103334583 on Build GRCh38 | Gain |
|
| nsv832253 | Chr.11:102975194 - 103170669 on Build GRCh38 | Gain |
|
| nsv556166 | Chr.11:103079240 - 103321803 on Build GRCh38 | Gain |
|
| esv3627547 | Chr.11:103066159 - 103156983 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map