Assay Details
Target Gene Details
Entrez Gene ID: | 728588 |
Gene Name: | membrane spanning 4-domains A18 |
Gene Aliases: |
- |
Location: |
Chr.11:60722909-60744213 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A18 | XM_011545420.2 | 1 | 944 | XP_011543722.1 |
| XM_011545421.2 | 1 | 946 | XP_011543723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051057 | Chr.11:60694023 - 60731810 on Build GRCh38 | Loss |
|
| nsv832175 | Chr.11:60579148 - 60749066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map