Assay Details
Target Gene Details
Entrez Gene ID: | 5954 |
Gene Name: | reticulocalbin 1 |
Gene Aliases: |
HEL-S-84, PIG20, RCAL, RCN |
Location: |
Chr.11:32090931-32105726 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCN1 | NM_002901.2 | NP_002892.1 | ||
| AK094360.1 | ||||
| AK129791.1 | ||||
| AY423728.1 | AAS00491.1 | |||
| BC010120.2 | AAH10120.1 | |||
| D42073.1 | BAA07670.1 | |||
| FJ224346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832107 | Chr.11:31987995 - 32167578 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map