Assay Details
Target Gene Details
Entrez Gene ID: | 5837 |
Gene Name: | phosphorylase, glycogen, muscle |
Gene Aliases: |
- |
Location: |
Chr.11:64746389-64760715 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYGM | NM_001164716.1 | NP_001158188.1 | ||
| AJ572752.1 | ||||
| AK056607.1 | ||||
| AK057547.1 | ||||
| AK299376.1 | ||||
| BM987822.1 | ||||
| X03031.1 | CAA26834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
|
| dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
|
| dgv153n27 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss |
|
| nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map