Assay Details
Target Gene Details
Entrez Gene ID: | 56935 |
Gene Name: | single-pass membrane protein with coiled-coil domains 4 |
Gene Aliases: |
C11orf75, FN5 |
Location: |
Chr.11:93478472-93586856 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMCO4 | NM_020179.2 | NP_064564.1 | ||
| XM_011542907.1 | XP_011541209.1 | |||
| XM_011542908.2 | XP_011541210.1 | |||
| XM_011542909.1 | XP_011541211.1 | |||
| XM_011542910.2 | XP_011541212.1 | |||
| XM_017018019.1 | XP_016873508.1 | |||
| AF197137.1 | AAF86046.1 | |||
| BC031564.1 | AAH31564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760208 | Chr.11:93439507 - 93533428 on Build GRCh38 | Loss |
|
| esv3627294 | Chr.11:93485306 - 93944317 on Build GRCh38 | Gain |
|
| dgv79n21 | Chr.11:93497971 - 93507681 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74485581] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map