Assay Details
Target Gene Details
Entrez Gene ID: | 54734 |
Gene Name: | RAB39A, member RAS oncogene family |
Gene Aliases: |
RAB39 |
Location: |
Chr.11:107928475-107963482 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB39A | NM_017516.2 | NP_059986.1 | ||
| AK292969.1 | ||||
| BC028064.1 | AAH28064.1 | |||
| HY107079.1 | ||||
| X99962.1 | CAA68227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750980 | Chr.11:107850215 - 108100259 on Build GRCh38 | Gain |
|
| esv3627655 | Chr.11:107740978 - 107953197 on Build GRCh38 | Gain |
|
| dgv1274n100 | Chr.11:107597955 - 108083284 on Build GRCh38 | Gain |
|
| esv3579902 | Chr.11:107921813 - 107940297 on Build GRCh38 | Loss |
|
| nsv1043139 | Chr.11:107854571 - 107998490 on Build GRCh38 | Gain |
|
| nsv521771 | Chr.11:107719609 - 108033920 on Build GRCh38 | Gain |
|
| dgv1276n100 | Chr.11:107693548 - 108059223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map