Assay Details
Target Gene Details
Entrez Gene ID: | 102724784 |
Gene Name: | uncharacterized LOC102724784 |
Gene Aliases: |
- |
Location: |
Chr.11:8768778-8810276 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724784 | NR_120590.1 | |||
| AI935056.1 | ||||
| AW593252.1 |
Target Gene Details
Entrez Gene ID: | 6764 |
Gene Name: | suppression of tumorigenicity 5 |
Gene Aliases: |
DENND2B, HTS1, p126 |
Location: |
Chr.11:8693352-8910951 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759803 | Chr.11:8763266 - 8988026 on Build GRCh38 | Gain |
|
| nsv553447 | Chr.11:8771075 - 8804419 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map