Assay Details
Target Gene Details
Entrez Gene ID: | 23201 |
Gene Name: | family with sequence similarity 168 member A |
Gene Aliases: |
KIAA0280, TCRP1 |
Location: |
Chr.11:73400478-73598189 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM168A | NM_001286050.1 | NP_001272979.1 | ||
| NM_001286051.1 | NP_001272980.1 | |||
| NM_015159.2 | NP_055974.1 | |||
| XM_017017415.1 | XP_016872904.1 | |||
| XM_017017416.1 | XP_016872905.1 | |||
| AK292054.1 | ||||
| BC052341.1 | AAH52341.1 | |||
| D87470.1 | ||||
| EF197985.1 | ||||
| EF363480.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs79296797] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map