Assay Details
Target Gene Details
Entrez Gene ID: | 100500938 |
Gene Name: | coiled-coil domain containing 179 |
Gene Aliases: |
- |
Location: |
Chr.11:22846922-22860971 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC179 | NM_001195637.1 | NP_001182566.1 | ||
| XM_011519802.2 | XP_011518104.1 | |||
| XM_011519803.2 | XP_011518105.1 | |||
| AI651461.1 |
Target Gene Details
Entrez Gene ID: | 2620 |
Gene Name: | growth arrest specific 2 |
Gene Aliases: |
- |
Location: |
Chr.11:22626113-22885930 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3625623 | Chr.11:22803579 - 22866492 on Build GRCh38 | Gain |
|
| nsv524283 | Chr.11:22821674 - 22859456 on Build GRCh38 | Loss |
|
| nsv1040326 | Chr.11:22841980 - 22872625 on Build GRCh38 | Loss |
|
| nsv1046773 | Chr.11:22597901 - 23427777 on Build GRCh38 | Loss |
|
| nsv1049802 | Chr.11:22813787 - 23248080 on Build GRCh38 | Gain |
|
| esv3625625 | Chr.11:22849447 - 22914921 on Build GRCh38 | Gain |
|
| nsv522782 | Chr.11:22847391 - 22869557 on Build GRCh38 | Loss |
|

Genomic Map