Assay Details
Target Gene Details
Entrez Gene ID: | 23201 |
Gene Name: | family with sequence similarity 168 member A |
Gene Aliases: |
KIAA0280, TCRP1 |
Location: |
Chr.11:73400478-73598189 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM168A | NM_001286050.1 | 7 | 810 | NP_001272979.1 |
| NM_001286051.1 | NP_001272980.1 | |||
| NM_015159.2 | 6 | 783 | NP_055974.1 | |
| XM_017017415.1 | 4 | 390 | XP_016872904.1 | |
| XM_017017416.1 | XP_016872905.1 | |||
| AK292054.1 | 6 | 673 | ||
| BC052341.1 | 6 | 777 | AAH52341.1 | |
| D87470.1 | 7 | 667 | ||
| EF197985.1 | ||||
| EF363480.1 | 6 | 777 |
More Information
Set Membership: |
|

Genomic Map