Assay Details
Target Gene Details
Entrez Gene ID: | 1072 |
Gene Name: | cofilin 1 |
Gene Aliases: |
CFL, HEL-S-15, cofilin |
Location: |
Chr.11:65854811-65858333 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFL1 | NM_005507.2 | NP_005498.1 | ||
| AK097690.1 | ||||
| AK303614.1 | ||||
| BC011005.2 | AAH11005.1 | |||
| BC012265.1 | AAH12265.1 | |||
| BC012318.1 | AAH12318.1 | |||
| BC018256.2 | AAH18256.1 | |||
| HM370399.1 | ||||
| X95404.1 | CAA64685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map