Assay Details
Target Gene Details
Entrez Gene ID: | 64231 |
Gene Name: | membrane spanning 4-domains A6A |
Gene Aliases: |
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090 |
Location: |
Chr.11:60171607-60184666 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832174 | Chr.11:60141794 - 60306435 on Build GRCh38 | Gain |
|
| nsv555150 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
| nsv524137 | Chr.11:60169506 - 60179779 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map