Assay Details
Target Gene Details
Entrez Gene ID: | 143458 |
Gene Name: | low density lipoprotein receptor class A domain containing 3 |
Gene Aliases: |
LRAD3 |
Location: |
Chr.11:35943981-36232136 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDLRAD3 | NM_001304263.1 | NP_001291192.1 | ||
| NM_001304264.1 | NP_001291193.1 | |||
| NM_174902.3 | NP_777562.1 | |||
| AK075546.1 | BAC11689.1 | |||
| AK293930.1 | ||||
| AK299186.1 | ||||
| BC042754.1 | ||||
| BC136286.1 | ||||
| BC143824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832122 | Chr.11:35912210 - 36082198 on Build GRCh38 | Gain |
|
| nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
|
| esv2422305 | Chr.11:35928148 - 36967365 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74422366] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map