Assay Details
Target Gene Details
Entrez Gene ID: | 338699 |
Gene Name: | ankyrin repeat domain 42 |
Gene Aliases: |
PPP1R79, SARP |
Location: |
Chr.11:83193739-83256099 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD42 | NM_001300972.1 | NP_001287901.1 | ||
| NM_001300973.1 | NP_001287902.1 | |||
| NM_001300975.1 | NP_001287904.1 | |||
| NM_001300976.1 | NP_001287905.1 | |||
| NM_001300977.1 | NP_001287906.1 | |||
| NM_182603.3 | NP_872409.2 | |||
| NR_125355.1 | ||||
| AK095193.1 | BAC04496.1 | |||
| AK310765.1 | ||||
| BC045621.1 | AAH45621.1 | |||
| BX648690.1 | ||||
| DQ508815.1 | ||||
| DQ508934.1 | ||||
| EF041819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33531 | Chr.11:82520885 - 83805659 on Build GRCh38 | Loss |
|

Genomic Map