Assay Details
Target Gene Details
Entrez Gene ID: | 2620 |
Gene Name: | growth arrest specific 2 |
Gene Aliases: |
- |
Location: |
Chr.11:22626113-22885930 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2 | XM_017017532.1 | XP_016873021.1 |
Target Gene Details
Entrez Gene ID: | 258010 |
Gene Name: | small VCP interacting protein |
Gene Aliases: |
- |
Location: |
Chr.11:22820263-22829862 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SVIP | NM_001320340.1 | NP_001307269.1 | ||
| NM_001320341.1 | NP_001307270.1 | |||
| NM_001320342.1 | NP_001307271.1 | |||
| NM_148893.2 | NP_683691.1 | |||
| NR_135213.1 | ||||
| AA347778.1 | ||||
| AF527534.1 | AAM88869.1 | |||
| AL833119.1 | ||||
| BF219509.1 | ||||
| BG531443.1 | ||||
| BI826588.1 | ||||
| DA306370.1 | ||||
| DB135755.1 | ||||
| DB466086.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523451 | Chr.11:22807060 - 22847391 on Build GRCh38 | Loss |
|
| esv3625623 | Chr.11:22803579 - 22866492 on Build GRCh38 | Gain |
|
| nsv524283 | Chr.11:22821674 - 22859456 on Build GRCh38 | Loss |
|
| nsv1046773 | Chr.11:22597901 - 23427777 on Build GRCh38 | Loss |
|
| nsv1049802 | Chr.11:22813787 - 23248080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map