Assay Details
Target Gene Details
Entrez Gene ID: | 84689 |
Gene Name: | membrane spanning 4-domains A14 |
Gene Aliases: |
MS4A16, NYD-SP21 |
Location: |
Chr.11:60396014-60417756 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A14 | NM_001079692.2 | NP_001073160.1 | ||
| NM_001261827.1 | NP_001248756.1 | |||
| NM_001261828.1 | NP_001248757.1 | |||
| NM_032597.4 | NP_115986.3 | |||
| NR_049731.1 | ||||
| NR_049732.1 | ||||
| NR_049733.1 | ||||
| NR_049734.1 | ||||
| AF367473.2 | AAK53409.2 | |||
| AK057418.1 | ||||
| AK314674.1 | ||||
| AY094610.1 | AAM48587.1 | |||
| AY094611.1 | AAM48588.1 | |||
| AY584610.1 | ||||
| BC107431.1 | AAI07432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744569 | Chr.11:60310239 - 60485227 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs137916808] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map