Assay Details
Target Gene Details
Entrez Gene ID: | 120406 |
Gene Name: | neurexophilin and PC-esterase domain family member 2 |
Gene Aliases: |
FAM55B |
Location: |
Chr.11:114464276-114813702 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE2 | XM_017017207.1 | XP_016872696.1 | ||
| XM_017017208.1 | XP_016872697.1 | |||
| XM_017017209.1 | XP_016872698.1 | |||
| XM_017017210.1 | XP_016872699.1 |
Target Gene Details
Entrez Gene ID: | 54827 |
Gene Name: | neurexophilin and PC-esterase domain family member 4 |
Gene Aliases: |
C11orf33, FAM55D |
Location: |
Chr.11:114570591-114596215 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NXPE4 | NM_001077639.1 | NP_001071107.1 | ||
| NM_017678.2 | NP_060148.2 | |||
| XM_011542881.2 | XP_011541183.1 | |||
| XM_011542882.2 | XP_011541184.1 | |||
| AK000134.1 | BAA90965.1 | |||
| AK225236.1 | ||||
| AY358805.1 | AAQ89165.1 | |||
| AY547307.1 | AAS55888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627794 | Chr.11:114568348 - 114603418 on Build GRCh38 | Gain |
|
| esv2759865 | Chr.11:114501563 - 114648403 on Build GRCh38 | Gain |
|
| nsv1053425 | Chr.11:114539109 - 114737646 on Build GRCh38 | Loss |
|
| nsv832274 | Chr.11:114535182 - 114685944 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map