Assay Details
Target Gene Details
Entrez Gene ID: | 50614 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 9 |
Gene Aliases: |
GALNAC-T9, GALNACT9 |
Location: |
Chr.12:132195829-132329364 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT9 | NM_001122636.1 | NP_001116108.1 | ||
| XM_017019371.1 | XP_016874860.1 | |||
| XM_017019372.1 | XP_016874861.1 | |||
| XM_017019373.1 | XP_016874862.1 | |||
| XM_017019374.1 | XP_016874863.1 | |||
| AB040672.2 | BAB13699.2 | |||
| AK054820.1 | ||||
| AK055773.1 | ||||
| DR001000.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560966 | Chr.12:132194518 - 132483313 on Build GRCh38 | Gain |
|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv520897 | Chr.12:132199317 - 132218324 on Build GRCh38 | Loss |
|
| nsv1041402 | Chr.12:132076412 - 132441297 on Build GRCh38 | Gain |
|
| nsv528119 | Chr.12:132174775 - 132218324 on Build GRCh38 | Loss |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
| dgv103n21 | Chr.12:132117051 - 132442096 on Build GRCh38 | Loss |
|
| nsv512300 | Chr.12:132211920 - 132214091 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs184380552] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map