Assay Details
Target Gene Details
Entrez Gene ID: | 23011 |
Gene Name: | RAB21, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.12:71754863-71793476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB21 | NM_014999.3 | NP_055814.1 | ||
| AF091035.1 | AAF00048.1 | |||
| BC021901.1 | AAH21901.1 | |||
| BC092475.1 | AAH92475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759907 | Chr.12:71708684 - 71914393 on Build GRCh38 | Gain |
|
| esv2658849 | Chr.12:71755874 - 71759207 on Build GRCh38 | Deletion |
|
| nsv559329 | Chr.12:71748682 - 71886804 on Build GRCh38 | Loss |
|
| nsv832451 | Chr.12:71584552 - 71806081 on Build GRCh38 | Loss |
|
| esv3629915 | Chr.12:71755848 - 71759299 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map