Assay Details
Target Gene Details
Entrez Gene ID: | 100506451 |
Gene Name: | RASSF8 atnisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:25954654-25959442 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF8-AS1 | NR_038227.1 | |||
| NR_038228.1 | ||||
| AK090817.1 | ||||
| BX109355.1 | ||||
| DA306887.1 | ||||
| DB074156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759886 | Chr.12:25839397 - 26008183 on Build GRCh38 | Loss |
|
| esv26656 | Chr.12:25955913 - 25961977 on Build GRCh38 | Gain+Loss |
|
| nsv557802 | Chr.12:25940228 - 25959225 on Build GRCh38 | Gain |
|

Genomic Map