Assay Details
Target Gene Details
Entrez Gene ID: | 11228 |
Gene Name: | Ras association domain family member 8 |
Gene Aliases: |
C12orf2, HOJ1 |
Location: |
Chr.12:25959031-26079892 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASSF8 | NM_001164748.1 | NP_001158220.1 | ||
| NM_007211.4 | NP_009142.2 | |||
| XM_017018746.1 | XP_016874235.1 | |||
| AB093206.1 | BAC98838.1 | |||
| AY665468.1 | AAV54603.1 | |||
| AY665469.1 | AAV54604.1 | |||
| AY665470.1 | AAV54605.1 | |||
| AY665471.1 | AAV54606.1 | |||
| BC030021.1 | AAH30021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759886 | Chr.12:25839397 - 26008183 on Build GRCh38 | Loss |
|
| esv26656 | Chr.12:25955913 - 25961977 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map