Assay Details
Target Gene Details
Entrez Gene ID: | 3892 |
Gene Name: | keratin 86 |
Gene Aliases: |
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX |
Location: |
Chr.12:52270507-52309163 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT86 | XM_005268866.4 | 1 | 495 | XP_005268923.1 |
| AK297794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759899 | Chr.12:52169480 - 52391735 on Build GRCh38 | Gain |
|
| dgv1501n100 | Chr.12:52237529 - 52281537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map