Assay Details
Target Gene Details
Entrez Gene ID: | 23141 |
Gene Name: | ankyrin repeat and LEM domain containing 2 |
Gene Aliases: |
KIAA0692, LEMD7, Lem4, MCPH16 |
Location: |
Chr.12:132725667-132761888 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKLE2 | NM_015114.2 | NP_055929.1 | ||
| XM_005266159.3 | XP_005266216.1 | |||
| XM_005266160.1 | XP_005266217.1 | |||
| XM_006719735.1 | XP_006719798.1 | |||
| XM_011534787.2 | XP_011533089.1 | |||
| XM_011534788.1 | XP_011533090.1 | |||
| XM_017019076.1 | XP_016874565.1 | |||
| AB014592.1 | BAA31667.1 | |||
| AK024061.1 | ||||
| BC001530.2 | AAH01530.2 | |||
| BC025347.1 | AAH25347.1 | |||
| BC043157.1 | AAH43157.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631312 | Chr.12:132735138 - 132794488 on Build GRCh38 | Loss |
|
| nsv561129 | Chr.12:132736267 - 132769626 on Build GRCh38 | Loss |
|
| nsv1047500 | Chr.12:132666737 - 132763400 on Build GRCh38 | Gain |
|
| nsv1037074 | Chr.12:132613063 - 132801953 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115110725,rs76962690] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map