Assay Details
Target Gene Details
Entrez Gene ID: | 160518 |
Gene Name: | DENN domain containing 5B |
Gene Aliases: |
- |
Location: |
Chr.12:31382223-31591018 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DENND5B | NM_001308339.1 | NP_001295268.1 | ||
| NM_144973.3 | NP_659410.3 | |||
| XM_005253326.2 | XP_005253383.1 | |||
| XM_005253327.3 | XP_005253384.1 | |||
| XM_005253330.3 | XP_005253387.1 | |||
| XM_006719040.3 | XP_006719103.1 | |||
| XM_011520590.2 | XP_011518892.1 | |||
| XM_017018896.1 | XP_016874385.1 | |||
| XM_017018897.1 | XP_016874386.1 | |||
| XM_017018898.1 | XP_016874387.1 | |||
| XM_017018899.1 | XP_016874388.1 | |||
| AB209341.1 | BAD92578.1 | |||
| AK095598.1 | BAC04583.1 | |||
| AK123642.1 | ||||
| AK125323.1 | BAC86129.1 | |||
| BX537924.1 | CAD97905.1 | |||
| CR749639.1 | CAH18433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751046 | Chr.12:31021299 - 31531799 on Build GRCh38 | Gain |
|
| nsv832364 | Chr.12:31315056 - 31495210 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map