Assay Details
Target Gene Details
Entrez Gene ID: | 319101 |
Gene Name: | keratin 73 |
Gene Aliases: |
IRT6IRS3, K6IRS3, KRT6IRS3 |
Location: |
Chr.12:52601813-52620133 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT73 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
| esv3629522 | Chr.12:52604253 - 52618660 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73320376] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map