Assay Details
Target Gene Details
Entrez Gene ID: | 50614 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 9 |
Gene Aliases: |
GALNAC-T9, GALNACT9 |
Location: |
Chr.12:132195829-132329364 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT9 | NM_001122636.1 | NP_001116108.1 | ||
| XM_017019372.1 | XP_016874861.1 | |||
| XM_017019373.1 | XP_016874862.1 | |||
| XM_017019375.1 | XP_016874864.1 | |||
| XM_017019376.1 | XP_016874865.1 | |||
| AB040672.2 | BAB13699.2 | |||
| AK055773.1 | ||||
| BC157845.1 | ||||
| BC171844.1 | ||||
| BM925654.1 | ||||
| DA206384.1 |
Target Gene Details
Entrez Gene ID: | 100130238 |
Gene Name: | uncharacterized LOC100130238 |
Gene Aliases: |
- |
Location: |
Chr.12:132275391-132280900 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130238 | NR_024563.1 | 4 | 2372 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560966 | Chr.12:132194518 - 132483313 on Build GRCh38 | Gain |
|
| nsv952512 | Chr.12:132192256 - 132711214 on Build GRCh38 | Deletion |
|
| nsv1041402 | Chr.12:132076412 - 132441297 on Build GRCh38 | Gain |
|
| nsv826563 | Chr.12:132125383 - 132622340 on Build GRCh38 | Gain |
|
| dgv103n21 | Chr.12:132117051 - 132442096 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74397849,rs77611298] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map